missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-chlorobenzoic acid, 98%

Manufacturer:  Alfa Aesar™ A1020106

 View more versions of this product

Catalog No. AAA1020106



Specifications

Specifications

2-Amino-4-chlorobenzoic acid
5g
C7H6ClNO2
743349
JYYLQSCZISREGY-UHFFFAOYSA-N
2-amino-4-chlorobenzoic acid
66646
98%
~232°C (decomposition)
89-77-0
MFCD00007778
4-chloroanthranilic acid, benzoic acid, 2-amino-4-chloro, 4-chloro-2-aminobenzoic acid, anthranilic acid, 4-chloro, 4-chloroanthranil acid, 4-chloro anthranilic acid, 2-carboxy-5-chloroaniline, unii-g55k85r12h, 2-amino-4-chloro-benzoic acid, 2,4-acba
C1=CC(=C(C=C1Cl)N)C(=O)O
171.58
171.58

Chemical Identifiers

89-77-0
171.58
JYYLQSCZISREGY-UHFFFAOYSA-N
66646
C1=CC(=C(C=C1Cl)N)C(=O)O
C7H6ClNO2
MFCD00007778
4-chloroanthranilic acid, benzoic acid, 2-amino-4-chloro, 4-chloro-2-aminobenzoic acid, anthranilic acid, 4-chloro, 4-chloroanthranil acid, 4-chloro anthranilic acid, 2-carboxy-5-chloroaniline, unii-g55k85r12h, 2-amino-4-chloro-benzoic acid, 2,4-acba
2-amino-4-chlorobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : DG1570000

SDS Documents