missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-chloro-6-methylpyrimidine, 98%, ACROS Organics™

Chemical Identifiers

CAS 5600-21-5
Molecular Formula C5H6ClN3
Molecular Weight (g/mol) 143.574
MDL Number MFCD00006091
InChI Key NPTGVVKPLWFPPX-UHFFFAOYSA-N
Synonym 2-amino-4-chloro-6-methylpyrimidine, 2-amino-6-chloro-4-methylpyrimidine, 2-pyrimidinamine, 4-chloro-6-methyl, 4-chloro-6-methyl-2-pyrimidinamine, 4-chloro-6-methylpyrimidin-2-ylamine, am inhibitor, am nitrification inhibitor, am pesticide, prepn. am, pyrimidine, 2-amino-4-chloro-6-methyl
PubChem CID 21810
ChEBI CHEBI:58960
IUPAC Name 4-chloro-6-methylpyrimidin-2-amine
SMILES CC1=CC(=NC(=N1)N)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC153750050
SDS View Documents
Thermo Fisher Scientific
AC153750050
5g Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

5600-21-5
143.574
NPTGVVKPLWFPPX-UHFFFAOYSA-N
21810
4-chloro-6-methylpyrimidin-2-amine
C5H6ClN3
MFCD00006091
2-amino-4-chloro-6-methylpyrimidine, 2-amino-6-chloro-4-methylpyrimidine, 2-pyrimidinamine, 4-chloro-6-methyl, 4-chloro-6-methyl-2-pyrimidinamine, 4-chloro-6-methylpyrimidin-2-ylamine, am inhibitor, am nitrification inhibitor, am pesticide, prepn. am, pyrimidine, 2-amino-4-chloro-6-methyl
CHEBI:58960
CC1=CC(=NC(=N1)N)Cl

Specifications

Authentic
White to Yellow
5g
97.5
98%
MFCD00006091
2-amino-4-chloro-6-methylpyrimidine, 2-amino-6-chloro-4-methylpyrimidine, 2-pyrimidinamine, 4-chloro-6-methyl, 4-chloro-6-methyl-2-pyrimidinamine, 4-chloro-6-methylpyrimidin-2-ylamine, am inhibitor, am nitrification inhibitor, am pesticide, prepn. am, pyrimidine, 2-amino-4-chloro-6-methyl
CC1=CC(=NC(=N1)N)Cl
143.574
CHEBI:58960
Needle-Like Crystalline Powder
2-Amino-4-chloro-6-methylpyrimidine, 98%
Glass Bottle
182°C to 186°C
5600-21-5
100.0
C5H6ClN3
24, 84
NPTGVVKPLWFPPX-UHFFFAOYSA-N
4-chloro-6-methylpyrimidin-2-amine
21810
143.58
≥97.5%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 227-018-3

RTECSNumber : UV6329600

TSCA : TSCA

SDS