Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-chloro-6-methoxypyrimidine, 99%

Chemical Identifiers

Chemical Name or Material 2-Amino-4-chloro-6-methoxypyrimidine
CAS 5734-64-5
Formula Weight 159.57
MDL Number MFCD00233535
Beilstein 127277
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6670322

 
alfa aesar™
H6670322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6670336

 
alfa aesar™
H6670336
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 168° to 173°C
Formula Weight 159.57
RTECS Number UV6260335
CAS 5734-64-5
Chemical Name or Material 2-Amino-4-chloro-6-methoxypyrimidine
Assay 99%
MDL Number MFCD00233535
TSCA TSCA
Beilstein 127277
Quantity 100g
Physical Form Solid