Please login to your online account to display your discounted pricing

2-Amino-4-chloro-6-methoxypyrimidine, 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  347090250

 View more versions of this product

Catalog No. AC347090250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 159.57
Quantity 25g
Color Light yellow
TSCA TSCA
Melting Point 169° to 176°C
RTECS Number UV6260335
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 2-Amino-4-chloro-6-methoxypyrimidine
Packaging Glass bottle
MDL Number MFCD00233535
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 95%
Assay Percent Range 95%
Molecular Formula C5H6ClN3O
CAS 5734-64-5

Specifications

Chemical Name or Material 2-Amino-4-chloro-6-methoxypyrimidine
Name Note 95%
CAS 5734-64-5
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C5H6ClN3O
Formula Weight 159.57
MDL Number MFCD00233535
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents