missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-chloro-6-methoxypyrimidine, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 347090050

Catalog No. AC347090050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Amino-4-chloro-6-methoxypyrimidine
5734-64-5
100.0
95%
C5H6ClN3O
5g
VFEYBTFCBZMBAU-UHFFFAOYSA-N
4-chloro-6-methoxypyrimidin-2-amine
79811
95%
95%
94.0
Authentic
Glass bottle
MFCD00233535
2-amino-4-chloro-6-methoxypyrimidine, 2-pyrimidinamine, 4-chloro-6-methoxy, 2-amino-6-chloro-4-methoxypyrimidine, pyrimidine, 2-amino-4-chloro-6-methoxy, 2-amino-4-methoxy-6-chloro pyrimidine, 4-chloro-6-methoxy-2-pyrimidinamine, 2-amino-4-chloro-6-methoxylpyrimidine, 2-amino-4-chloro-6-methoxy pyrimidine, 2-amino-4-chloro-6-methoxy-pyrimidine, 4-chloro-6-methoxy-pyrimidin-2-ylamine
COC1=CC(Cl)=NC(N)=N1
159.57
159.57
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

RTECSNumber : UV6260335

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only