Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-chloro-6-isopropylpyrimidine, 97%

$146.77 - $415.86

Chemical Identifiers

Chemical Name or Material 2-Amino-4-chloro-6-isopropylpyrimidine
CAS 73576-33-7
Molecular Formula C7H10ClN3
Formula Weight 171.63
MDL Number MFCD03840198
Synonym 4-Chloro-6-isopropyl-2-pyrimidinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2599703

  SDS
alfa aesar™
H2599703
1g Each for $415.86

AAH25997MD

  SDS
alfa aesar™
H25997MD
250mg Each for $146.77
Description & Specifications

Specifications

Formula Weight 171.63
Melting Point 98° to 101°C
CAS 73576-33-7
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD03840198
Synonym 4-Chloro-6-isopropyl-2-pyrimidinamine
Health Hazard 1 Warning
Chemical Name or Material 2-Amino-4-chloro-6-isopropylpyrimidine
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Molecular Formula C7H10ClN3
Assay 97%

Applications
2-Amino-4-chloro-6-isopropylpyrimidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.