Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-chloro-6-(4-morpholinyl)pyrimidine, 99%

$119.45 - $412.53

Chemical Identifiers

Chemical Name or Material 2-Amino-4-chloro-6-(4-morpholinyl)pyrimidine
CAS 339016-18-1
Molecular Formula C8H11ClN4O
Formula Weight 214.66
MDL Number MFCD00664697
Synonym 4-Chloro-6-(4-morpholinyl)-2-pyrimidinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH50538MD

 
alfa aesar™
H50538MD
250mg Each for $119.45

AAH5053803

 
alfa aesar™
H5053803
1g Each for $412.53
Description & Specifications

Specifications

Formula Weight 214.66
Chemical Name or Material 2-Amino-4-chloro-6-(4-morpholinyl)pyrimidine
Melting Point 212° to 214°C
Quantity 250mg
Assay 99%
Molecular Formula C8H11ClN4O
MDL Number MFCD00664697
CAS 339016-18-1
Synonym 4-Chloro-6-(4-morpholinyl)-2-pyrimidinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.