Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-chloro-5-methylbenzenesulfonic acid, 97%

$67.58 - $261.68

Chemical Identifiers

Chemical Name or Material 2-Amino-4-chloro-5-methylbenzenesulfonic acid
CAS 88-51-7
Molecular Formula C7H8ClNO3S
Formula Weight 221.66
MDL Number MFCD00035771
Synonym 6-Amino-4-chloro-m-toluenesulfonic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1919809

  SDS
alfa aesar™
A1919809
10g Each for $67.58

AAA1919818

  SDS
alfa aesar™
A1919818
50g Each for $261.68
Description & Specifications

Specifications

Formula Weight 221.66
Chemical Name or Material 2-Amino-4-chloro-5-methylbenzenesulfonic acid
TSCA TSCA
Quantity 10g
Assay 97%
Molecular Formula C7H8ClNO3S
MDL Number MFCD00035771
CAS 88-51-7
Synonym 6-Amino-4-chloro-m-toluenesulfonic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number XT6330000
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.