missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-chloro-5,6-dimethylpyrimidine, 97%

Manufacturer:  Alfa Aesar™ H54552MD

 View more versions of this product

Catalog No. AAH54552MD



Description

Description

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in Chloroform and Ethanol

Notes
Store at -20°C. Keep away from light and heat.
Specifications

Specifications

2-Amino-4-chloro-5,6-dimethylpyrimidine
14394-61-7
MFCD00462111
Soluble in Chloroform and Ethanol
CC1=C(N=C(N=C1Cl)N)C
157.601
157.6
250mg
C6H8ClN3
2-amino-4-chloro-5,6-dimethyl-pyrimidine
RRMVDLLCVGZDMH-UHFFFAOYSA-N
4-chloro-5,6-dimethylpyrimidin-2-amine
248655
97%

Chemical Identifiers

14394-61-7
157.601
RRMVDLLCVGZDMH-UHFFFAOYSA-N
248655
CC1=C(N=C(N=C1Cl)N)C
C6H8ClN3
MFCD00462111
2-amino-4-chloro-5,6-dimethyl-pyrimidine
4-chloro-5,6-dimethylpyrimidin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents