missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-bromopyrimidine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6435803

 View more versions of this product

Catalog No. AAH6435803


Description

Description

It is used as a pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate. It is also an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

2-Amino-4-bromopyrimidine
White
MFCD09999168
111375
Slightly soluble in water.
C1=CN=C(N=C1Br)N
174.001
174
Powder
343926-69-2
C4H4BrN3
1 g
2-amino-4-bromopyrimidine, 4-bromo-2-pyrimidinamine, 2-pyrimidinamine,4-bromo, 4-bromanylpyrimidin-2-amine, 4-bromopyrimidine-2-ylamine, 2-amino-6-bromopyrimidine, 4-bromo-2-aminopyrimidine, 2-pyrimidinamine,4-bromo-9ci, 2-pyrimidinamine, 4-bromo, 4-bromo-pyrimidin-2-ylamine
MINURGGJJKMQQQ-UHFFFAOYSA-N
4-bromopyrimidin-2-amine
12803496
98%

Chemical Identifiers

343926-69-2
174.001
MINURGGJJKMQQQ-UHFFFAOYSA-N
12803496
C1=CN=C(N=C1Br)N
C4H4BrN3
MFCD09999168
2-amino-4-bromopyrimidine, 4-bromo-2-pyrimidinamine, 2-pyrimidinamine,4-bromo, 4-bromanylpyrimidin-2-amine, 4-bromopyrimidine-2-ylamine, 2-amino-6-bromopyrimidine, 4-bromo-2-aminopyrimidine, 2-pyrimidinamine,4-bromo-9ci, 2-pyrimidinamine, 4-bromo, 4-bromo-pyrimidin-2-ylamine
4-bromopyrimidin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only