Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-bromopyridine, 97%

$83.58 - $230.89

Chemical Identifiers

Chemical Name or Material 2-Amino-4-bromopyridine
CAS 84249-14-9
Molecular Formula C5H5BrN2
Formula Weight 173.01
MDL Number MFCD01646115
Synonym 4-Bromo-2-pyridinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3150003

  SDS
alfa aesar™
H3150003
1g Each for $230.89

AAH31500MD

  SDS
alfa aesar™
H31500MD
250mg Each for $83.58
Description & Specifications

Specifications

Formula Weight 173.01
Chemical Name or Material 2-Amino-4-bromopyridine
Assay 97%
Melting Point 144° to 147°C
Molecular Formula C5H5BrN2
Quantity 1g
MDL Number MFCD01646115
CAS 84249-14-9
Synonym 4-Bromo-2-pyridinamine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.