missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-bromophenol, 98%

$31.05 - $321.07

Chemical Identifiers

CAS 40925-68-6
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
MDL Number MFCD00235171
InChI Key JHRIPENGTGSNPJ-UHFFFAOYSA-N
Synonym 2-amino-4-bromo-phenol
PubChem CID 351840
IUPAC Name 2-amino-4-bromophenol
SMILES C1=CC(=C(C=C1Br)N)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55763MD View Documents Alfa Aesar™
H55763MD
250mg Each for $31.05
 
AAH5576303 View Documents Alfa Aesar™
H5576303
1g Each for $84.64
 
AAH5576306 View Documents Alfa Aesar™
H5576306
5g Each for $321.07
 
Specifications

Chemical Identifiers

40925-68-6
188.024
JHRIPENGTGSNPJ-UHFFFAOYSA-N
351840
C1=CC(=C(C=C1Br)N)O
C6H6BrNO
MFCD00235171
2-amino-4-bromo-phenol
2-amino-4-bromophenol

Specifications

130°C to 135°C
40925-68-6
MFCD00235171
2-amino-4-bromo-phenol
C1=CC(=C(C=C1Br)N)O
188.024
188.02
2-Amino-4-bromophenol
250mg
C6H6BrNO
3236448
JHRIPENGTGSNPJ-UHFFFAOYSA-N
2-amino-4-bromophenol
351840
98%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H334-H315-H319-H317-H335
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319

TSCA : No

Recommended Storage : Ambient temperatures