missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-bromophenol, 98%

Manufacturer:  Alfa Aesar™ H5576303

 View more versions of this product

Catalog No. AAH5576303



Specifications

Specifications

2-Amino-4-bromophenol
1g
C6H6BrNO
3236448
JHRIPENGTGSNPJ-UHFFFAOYSA-N
2-amino-4-bromophenol
351840
98%
130°C to 135°C
40925-68-6
MFCD00235171
2-amino-4-bromo-phenol, 2-amino-4-bromophenole, 2-amino-4bromophenol, 2-amino4-bromophenol, 4-bromo-2-aminophenol, 5-bromo-2-hydroxyaniline, acmc-209jho, ksc493s8p, phenol, 2-amino-4-bromo, pubchem4511
C1=CC(=C(C=C1Br)N)O
188.024
188.02

Chemical Identifiers

40925-68-6
188.024
JHRIPENGTGSNPJ-UHFFFAOYSA-N
351840
C1=CC(=C(C=C1Br)N)O
C6H6BrNO
MFCD00235171
2-amino-4-bromo-phenol, 2-amino-4-bromophenole, 2-amino-4bromophenol, 2-amino4-bromophenol, 4-bromo-2-aminophenol, 5-bromo-2-hydroxyaniline, acmc-209jho, ksc493s8p, phenol, 2-amino-4-bromo, pubchem4511
2-amino-4-bromophenol
Safety and Handling

Safety and Handling

GHS H Statement
H301-H334-H315-H319-H317-H335
Toxic if swallowed.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

Documents