missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-4-bromobenzoic acid, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H55829MD
Description
2-Amino-4-bromobenzoic acid, , is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-4-bromobenzoic acid, , is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water (slightly).
Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
2-Amino-4-bromobenzoic acid | |
230°C to 234°C | |
MFCD03618454 | |
4-bromoanthranilic acid, benzoic acid, 2-amino-4-bromo, 5-bromo-2-carboxyaniline, 2-amino-4-bromo-benzoic acid, 2-amino-4-brombenzoic acid, zlchem 336, pubchem13462, acmc-1cb3t, ksc207e6b, pharmabridge p-1344 | |
BNNICQAVXPXQAH-UHFFFAOYSA-N | |
2-amino-4-bromobenzoic acid | |
88691 | |
97% |
20776-50-5 | |
C7H6BrNO2 | |
250 mg | |
Soluble in water (slightly). | |
C1=CC(=C(C=C1Br)N)C(=O)O | |
216.034 | |
216.03 |
Chemical Identifiers
20776-50-5 | |
216.034 | |
BNNICQAVXPXQAH-UHFFFAOYSA-N | |
88691 | |
C1=CC(=C(C=C1Br)N)C(=O)O |
C7H6BrNO2 | |
MFCD03618454 | |
4-bromoanthranilic acid, benzoic acid, 2-amino-4-bromo, 5-bromo-2-carboxyaniline, 2-amino-4-bromo-benzoic acid, 2-amino-4-brombenzoic acid, zlchem 336, pubchem13462, acmc-1cb3t, ksc207e6b, pharmabridge p-1344 | |
2-amino-4-bromobenzoic acid |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 244-025-7
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only