Learn More
2-Amino-4,6-dimethylpyrimidine, 98%, ACROS Organics™
$155.41 - $155.41
Chemical Identifiers
CAS | 767-15-7 |
---|---|
Molecular Formula | C6H9N3 |
Molecular Weight (g/mol) | 123.16 |
MDL Number | MFCD00006102 |
InChI Key | IDQNBVFPZMCDDN-UHFFFAOYSA-N |
Synonym | 2-amine-4,6-dimethylpyrimidine, 2-amino-4,6-dimethyl pyrimidine, 2-amino-4,6-dimethylpyrimidine, 2-pyrimidinamine, 4,6-dimethyl, 4,6-dimethyl-2-aminopyrimidine, 4,6-dimethyl-2-pyrimidinamine, 4,6-dimethylpyrimidin-2-ylamine, acetylacetoneguanidine, pyrimidine, 2-amino-4,6-dimethyl, unii-4s48mi253m |
PubChem CID | 13021 |
IUPAC Name | 4,6-dimethylpyrimidin-2-amine |
SMILES | CC1=CC(=NC(=N1)N)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC103590250
|
Thermo Fisher Scientific
AC103590250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC103591000
|
Thermo Fisher Scientific
103591000 |
100g | Glass bottle |
Each for $155.41
|
|
Chemical Identifiers
767-15-7 | |
123.16 | |
IDQNBVFPZMCDDN-UHFFFAOYSA-N | |
13021 | |
CC1=CC(=NC(=N1)N)C |
C6H9N3 | |
MFCD00006102 | |
2-amine-4,6-dimethylpyrimidine, 2-amino-4,6-dimethyl pyrimidine, 2-amino-4,6-dimethylpyrimidine, 2-pyrimidinamine, 4,6-dimethyl, 4,6-dimethyl-2-aminopyrimidine, 4,6-dimethyl-2-pyrimidinamine, 4,6-dimethylpyrimidin-2-ylamine, acetylacetoneguanidine, pyrimidine, 2-amino-4,6-dimethyl, unii-4s48mi253m | |
4,6-dimethylpyrimidin-2-amine |
Specifications
Authentic | |
White to Beige | |
25g | |
97.5 | |
98% | |
MFCD00006102 | |
IDQNBVFPZMCDDN-UHFFFAOYSA-N | |
4,6-dimethylpyrimidin-2-amine | |
13021 | |
Crystalline Powder | |
2-Amino-4, 6-dimethylpyrimidine, 98% |
Glass bottle | |
150°C to 154°C | |
767-15-7 | |
100.0 | |
C6H9N3 | |
2-amine-4,6-dimethylpyrimidine, 2-amino-4,6-dimethyl pyrimidine, 2-amino-4,6-dimethylpyrimidine, 2-pyrimidinamine, 4,6-dimethyl, 4,6-dimethyl-2-aminopyrimidine, 4,6-dimethyl-2-pyrimidinamine, 4,6-dimethylpyrimidin-2-ylamine, acetylacetoneguanidine, pyrimidine, 2-amino-4,6-dimethyl, unii-4s48mi253m | |
CC1=CC(=NC(=N1)N)C | |
123.16 | |
123.16 | |
98% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 212-181-5
RTECSNumber : UV6260496
TSCA : TSCA