missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4,6-dimethylpyrimidine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC103590250

 View more versions of this product

Catalog No. AC103590250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Amino-4, 6-dimethylpyrimidine
Glass bottle
150°C to 154°C
98%
97.5
98%
MFCD00006102
IDQNBVFPZMCDDN-UHFFFAOYSA-N
4,6-dimethylpyrimidin-2-amine
13021
Crystalline Powder
Authentic
White to Beige
25g
767-15-7
100.0
C6H9N3
2-amine-4,6-dimethylpyrimidine, 2-amino-4,6-dimethyl pyrimidine, 2-amino-4,6-dimethylpyrimidine, 2-pyrimidinamine, 4,6-dimethyl, 4,6-dimethyl-2-aminopyrimidine, 4,6-dimethyl-2-pyrimidinamine, 4,6-dimethylpyrimidin-2-ylamine, acetylacetoneguanidine, pyrimidine, 2-amino-4,6-dimethyl, unii-4s48mi253m
CC1=CC(=NC(=N1)N)C
123.16
123.16
98%

Chemical Identifiers

767-15-7
123.16
IDQNBVFPZMCDDN-UHFFFAOYSA-N
13021
CC1=CC(=NC(=N1)N)C
C6H9N3
MFCD00006102
2-amine-4,6-dimethylpyrimidine, 2-amino-4,6-dimethyl pyrimidine, 2-amino-4,6-dimethylpyrimidine, 2-pyrimidinamine, 4,6-dimethyl, 4,6-dimethyl-2-aminopyrimidine, 4,6-dimethyl-2-pyrimidinamine, 4,6-dimethylpyrimidin-2-ylamine, acetylacetoneguanidine, pyrimidine, 2-amino-4,6-dimethyl, unii-4s48mi253m
4,6-dimethylpyrimidin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-181-5

RTECSNumber : UV6260496

TSCA : TSCA

SDS Documents