missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4,6-dimethylpyridine, 99%, ACROS Organics™

Manufacturer:  Acros Organics AC103580250

 View more versions of this product

Catalog No. AC103580250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Amino-4, 6-dimethylpyridine
Authentic
235.0°C
65.0°C to 69.5°C
5407-87-4
100.0
C7H10N2
2,4-lutidine, 6-amino
BRBUBVKGJRPRRD-UHFFFAOYSA-N
4,6-dimethylpyridin-2-amine
21507
Crystalline Powder
121°C
Glass bottle
White to Brown
25g
98.5
99%
(CH3)2(C5H2N)NH2
Solubility in water: 5.5 g/100mL (20 c).
CC1=CC(=NC(=C1)N)C
122.171
122.17
99%

Chemical Identifiers

5407-87-4
122.171
2,4-lutidine, 6-amino
4,6-dimethylpyridin-2-amine
C7H10N2
BRBUBVKGJRPRRD-UHFFFAOYSA-N
21507
CC1=CC(=NC(=C1)N)C
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents