Please login to your online account to display your discounted pricing

2-Amino-4,6-dimethylpyridine, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Amino-4, 6-dimethylpyridine
CAS 5407-87-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H10N2
Linear Formula (CH3)2(C5H2N)NH2
Formula Weight 122.17
Synonym 6-Amino-2, 4-lutidine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC103580250

  SDS
thermo fisher scientific
AC103580250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC103580050

  SDS
thermo fisher scientific
AC103580050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 122.17
Quantity 25g
Color White to light brown
Melting Point 65° to 70°C
CAS Max % 100.0
Linear Formula (CH3)2(C5H2N)NH2
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay Percent Range 99%
Molecular Formula C7H10N2
Chemical Name or Material 2-Amino-4, 6-dimethylpyridine
CAS 5407-87-4
CAS Min % 98.5
Infrared Spectrum Authentic
Synonym 6-Amino-2, 4-lutidine
Packaging Glass bottle
Safety and Handling

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.