Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4,6-dimethoxypyrimidine, 98%

$36.95 - $85.58

Chemical Identifiers

Chemical Name or Material 2-Amino-4,6-dimethoxypyrimidine
CAS 36315-01-2
Molecular Formula C6H9N3O2
Formula Weight 155.16
MDL Number MFCD00038832
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2015209

 
alfa aesar™
B2015209
10g Each for $85.58

AAB2015288

 
alfa aesar™
B2015288
2.5g Each for $36.95
Description & Specifications

Specifications

Formula Weight 155.16
Chemical Name or Material 2-Amino-4,6-dimethoxypyrimidine
TSCA TSCA
Melting Point 94° to 96°C
Assay 98%
Quantity 10g
Molecular Formula C6H9N3O2
MDL Number MFCD00038832
CAS 36315-01-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.