Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4,6-dihydroxypyrimidine, 98%

$55.88 - $178.55

Chemical Identifiers

Chemical Name or Material 2-Amino-4,6-dihydroxypyrimidine
CAS 56-09-7
Molecular Formula C4H5N3O2
Formula Weight 127.1
MDL Number MFCD00006094
Beilstein 510297
Synonym 2-Aminopyrimidine-4,6-diol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1181314

 
alfa aesar™
A1181314
25g Each for $55.88

AAA1181322

 
alfa aesar™
A1181322
100g Each for $178.55
Description & Specifications

Specifications

Formula Weight 127.1
Chemical Name or Material 2-Amino-4,6-dihydroxypyrimidine
TSCA TSCA
Melting Point >300°C
Assay 98%
Quantity 25g
Molecular Formula C4H5N3O2
MDL Number MFCD00006094
CAS 56-09-7
Beilstein 510297
Synonym 2-Aminopyrimidine-4,6-diol
RTECS Number UW7361000

Applications
2-Amino-4,6-dihydroxypyrimidine acts as an intermediate in the production of antimicrobial guanylsulfonamides. It also acts as an intermediate in pharmaceuticals and agrochemicals.

Solubility
Soluble in Aqueous Alkali.

Notes
Store away from oxidizing agents. Store at -20°C. Store in a cool, dry, well-ventilated area.