missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-4,6-dihydroxypyrimidine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1181322
Description
2-Amino-4,6-dihydroxypyrimidine acts as an intermediate in the production of antimicrobial guanylsulfonamides. It also acts as an intermediate in pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-4,6-dihydroxypyrimidine acts as an intermediate in the production of antimicrobial guanylsulfonamides. It also acts as an intermediate in pharmaceuticals and agrochemicals.
Solubility
Soluble in Aqueous Alkali.
Notes
Store away from oxidizing agents. Store at -20°C. Store in a cool, dry, well-ventilated area.
Specifications
2-Amino-4,6-dihydroxypyrimidine | |
>300°C | |
MFCD00006094 | |
510297 | |
Soluble in Aqueous Alkali. | |
NC1=NC(O)=CC(=O)N1 | |
127.10 | |
127.1 |
56-09-7 | |
C4H5N3O2 | |
100 g | |
2-amino-4,6-dihydroxypyrimidine, 2-aminopyrimidine-4,6-diol, 4 1h-pyrimidinone, 2-amino-6-hydroxy, 2-amino-6-hydroxypyrimidin-4 3h-one, 2-amino-4,6-dioxypyrimidine, 2-amino-4,6-pyrimidinedione, 4,6-dihydroxy-2-aminopyrimidine, 2-amino-4,6-pyrimidinediol, 2-amino-6-hydroxy-1h-pyrimidin-4-one, 2-amino-6-hydroxy-4 1h-pyrimidinone | |
AUFJTVGCSJNQIF-UHFFFAOYSA-N | |
2-amino-4-hydroxy-1H-pyrimidin-6-one | |
66131 | |
98% |
Chemical Identifiers
56-09-7 | |
127.10 | |
AUFJTVGCSJNQIF-UHFFFAOYSA-N | |
66131 | |
NC1=NC(O)=CC(=O)N1 |
C4H5N3O2 | |
MFCD00006094 | |
2-amino-4,6-dihydroxypyrimidine, 2-aminopyrimidine-4,6-diol, 4 1h-pyrimidinone, 2-amino-6-hydroxy, 2-amino-6-hydroxypyrimidin-4 3h-one, 2-amino-4,6-dioxypyrimidine, 2-amino-4,6-pyrimidinedione, 4,6-dihydroxy-2-aminopyrimidine, 2-amino-4,6-pyrimidinediol, 2-amino-6-hydroxy-1h-pyrimidin-4-one, 2-amino-6-hydroxy-4 1h-pyrimidinone | |
2-amino-4-hydroxy-1H-pyrimidin-6-one |
Safety and Handling
EINECSNumber : 200-256-5
RTECSNumber : UW7361000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only