missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4,6-difluorobenzothiazole, 97%

Manufacturer:  Alfa Aesar™ H5550606

 View more versions of this product

Catalog No. AAH5550606



Description

Description

Applications
2-Amino-4,6-difluorobenzothiazole is used as drugs, food, cosmetics

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2-Amino-4,6-difluorobenzothiazole
216°C to 220°C
119256-40-5
MFCD00276099
DDKKXSCVPKDRRS-UHFFFAOYSA-N
4,6-difluoro-1,3-benzothiazol-2-amine
737406
Powder
Brown to White
5g
C7H4F2N2S
2-amino-4,6-difluoro-1,3-benzothiazol
C1=C(C=C2C(=C1F)N=C(S2)N)F
186.18
186.18
97%

Chemical Identifiers

119256-40-5
186.18
DDKKXSCVPKDRRS-UHFFFAOYSA-N
737406
C1=C(C=C2C(=C1F)N=C(S2)N)F
C7H4F2N2S
MFCD00276099
2-amino-4,6-difluoro-1,3-benzothiazol
4,6-difluoro-1,3-benzothiazol-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents