Learn More
2-Amino-4,6-dichloropyrimidine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1133006
Description
Microwave-assisted synthesis of pyrazolo [3, 4-d] pyrimidines from 2-amino-4, 6-dichloropyrimidine-5-carbaldehyde is under solvent-free conditions. A series of 6-alkynyl-2,4-diaminopyrimidine derivatives bearing various substituents at alkynyl moiety was prepared by the Sonogashira cross-coupling reaction of 2,4-diamino-6-iodopyrimidine using Pd(PPh3)2Cl2 as catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMicrowave-assisted synthesis of pyrazolo [3, 4-d] pyrimidines from 2-amino-4, 6-dichloropyrimidine-5-carbaldehyde is under solvent-free conditions. A series of 6-alkynyl-2,4-diaminopyrimidine derivatives bearing various substituents at alkynyl moiety was prepared by the Sonogashira cross-coupling reaction of 2,4-diamino-6-iodopyrimidine using Pd(PPh3)2Cl2 as catalyst.
Solubility
Insoluble in water, soluble in acetone, two monomer (VCM) and thermal toluene.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
2-Amino-4,6-dichloropyrimidine | |
218°C to 222°C | |
MFCD00006090 | |
118454 | |
Insoluble in water,soluble in acetone,two monomer (VCM) and thermal toluene. | |
C1=C(N=C(N=C1Cl)N)Cl | |
163.989 | |
164 |
56-05-3 | |
C4H3Cl2N3 | |
5 g | |
2-amino-4,6-dichloropyrimidine, 4,6-dichloro-2-pyrimidinamine, 2-pyrimidinamine, 4,6-dichloro, 4,6-dichloropyrimidin-2-ylamine, ccris 7460, 4,6-dichloro-pyrimidin-2-ylamine, pyrimidine, 2-amino-4,6-dichloro, 2-amino-4,6-dichloro pyrimidine, 4,6-dichloropyrimidine-2-ylamine, zlchem 351 | |
JPZOAVGMSDSWSW-UHFFFAOYSA-N | |
4,6-dichloropyrimidin-2-amine | |
65522 | |
98% |
Chemical Identifiers
56-05-3 | |
163.989 | |
JPZOAVGMSDSWSW-UHFFFAOYSA-N | |
65522 | |
C1=C(N=C(N=C1Cl)N)Cl |
C4H3Cl2N3 | |
MFCD00006090 | |
2-amino-4,6-dichloropyrimidine, 4,6-dichloro-2-pyrimidinamine, 2-pyrimidinamine, 4,6-dichloro, 4,6-dichloropyrimidin-2-ylamine, ccris 7460, 4,6-dichloro-pyrimidin-2-ylamine, pyrimidine, 2-amino-4,6-dichloro, 2-amino-4,6-dichloro pyrimidine, 4,6-dichloropyrimidine-2-ylamine, zlchem 351 | |
4,6-dichloropyrimidin-2-amine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 200-253-9
RTECSNumber : UV6260485
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only