missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4,6-dichloropyrimidine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC103492500

 View more versions of this product

Catalog No. AC103492500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Amino-4, 6-dichloropyrimidine
94.0
Authentic
216° to 226°C
Liquid
250g
C4H3Cl2N3
MFCD00006090
56-05-3
100.0
Clear colorless to light yellow
Technical Grade
≥94% (HPLC)
95%
163.99
24, 81

Chemical Identifiers

56-05-3
100.0
C4H3Cl2N3
MFCD00006090
≥94% (HPLC)
94.0
95%
163.99
2-Amino-4, 6-dichloropyrimidine
Technical Grade