Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4,6-dichloro-5-methylphenol, 98%

$203.19 - $680.36

Chemical Identifiers

Chemical Name or Material 2-Amino-4,6-dichloro-5-methylphenol
CAS 40677-44-9
Molecular Formula C7H7Cl2NO
Formula Weight 192.04
MDL Number MFCD00071776
Synonym 6-Amino-2,4-dichloro-m-cresol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1854736

 
alfa aesar™
A1854736
500g Each for $680.36

AAA1854722

 
alfa aesar™
A1854722
100g Each for $203.19
Description & Specifications

Specifications

Formula Weight 192.04
Chemical Name or Material 2-Amino-4,6-dichloro-5-methylphenol
TSCA TSCA
Assay 98%
Quantity 500g
Molecular Formula C7H7Cl2NO
MDL Number MFCD00071776
CAS 40677-44-9
Synonym 6-Amino-2,4-dichloro-m-cresol
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.