Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Amino-4',5'-methylenedioxyacetophenone hydrochloride, 98%

$290.81 - $911.59

Chemical Identifiers

Chemical Name or Material 2'-Amino-4',5'-methylenedioxyacetophenone hydrochloride
CAS 93983-01-8
Molecular Formula C9H9NO3·HCl
Formula Weight 215.64
MDL Number MFCD00053022
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2178814

  SDS
alfa aesar™
B2178814
25g Each for $290.81

AAB2178822

  SDS
alfa aesar™
B2178822
100g Each for $911.59
Description & Specifications

Specifications

Formula Weight 215.64
Chemical Name or Material 2'-Amino-4',5'-methylenedioxyacetophenone hydrochloride
MDL Number MFCD00053022
Assay 98%
Molecular Formula C9H9NO3·HCl
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 93983-01-8
Quantity 25g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.