Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Amino-4',5'-methylenedioxyacetophenone, 98%

$161.01 - $161.01

Chemical Identifiers

Chemical Name or Material 2'-Amino-4',5'-methylenedioxyacetophenone
CAS 28657-75-2
Molecular Formula C9H9NO3
Formula Weight 179.17g/mol
MDL Number MFCD00075855
Synonym 5-Acetyl-6-amino-1,3-benzodioxole
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AAA1677109

 
alfa aesar™
A16771-09
Each for $161.01
Description & Specifications

Specifications

Melting Point 170° to 174°C
Quantity 10g
Chemical Name or Material 2'-Amino-4',5'-methylenedioxyacetophenone
CAS 28657-75-2
Assay 98%
Molecular Formula C9H9NO3
Formula Weight 179.17g/mol
MDL Number MFCD00075855
Synonym 5-Acetyl-6-amino-1,3-benzodioxole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.