Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4,5-dimethylthiazole hydrobromide, 98%

$45.81 - $127.18

Chemical Identifiers

Chemical Name or Material 2-Amino-4,5-dimethylthiazole hydrobromide
CAS 7170-76-5
Molecular Formula C5H8N2S·HBr
Formula Weight 209.11
MDL Number MFCD00035212
Beilstein 3690705
Synonym 4,5-Dimethyl-2-thiazolamine hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2080306

  SDS
alfa aesar™
B2080306
5g Each for $45.81

AAB2080314

  SDS
alfa aesar™
B2080314
25g Each for $127.18
Description & Specifications

Specifications

Formula Weight 209.11
Chemical Name or Material 2-Amino-4,5-dimethylthiazole hydrobromide
TSCA TSCA
Melting Point 287° to 293°C
Assay 98%
Quantity 5g
Molecular Formula C5H8N2S·HBr
MDL Number MFCD00035212
CAS 7170-76-5
Beilstein 3690705
Synonym 4,5-Dimethyl-2-thiazolamine hydrobromide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.