Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4,5-dimethoxybenzonitrile, 98%

Chemical Identifiers

Chemical Name or Material 2-Amino-4,5-dimethoxybenzonitrile
CAS 26961-27-3
Molecular Formula C9H10N2O2
Formula Weight 178.19
MDL Number MFCD00058682
UN Number UN3439
Beilstein 2372149
Synonym 4-Amino-5-cyanoveratrole; 2-Cyano-4,5-dimethoxyaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0954506

  SDS
alfa aesar™
L0954506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0954514

  SDS
alfa aesar™
L0954514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 178.19
Molecular Formula C9H10N2O2
MDL Number MFCD00058682
Chemical Name or Material 2-Amino-4,5-dimethoxybenzonitrile
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
UN Number UN3439
CAS 26961-27-3
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Assay 98%
Beilstein 2372149
Melting Point 97° to 101°C
Synonym 4-Amino-5-cyanoveratrole; 2-Cyano-4,5-dimethoxyaniline
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.