missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4,5-dimethoxybenzonitrile, 98%

Manufacturer:  Alfa Aesar™ L0954514

 View more versions of this product

Catalog No. AAL0954514



Specifications

Specifications

2-Amino-4,5-dimethoxybenzonitrile
25g
C9H10N2O2
UN3439
6-aminoveratronitrile, 2-cyano-4,5-dimethoxyaniline, 4-amino-5-cyanoveratrole, 6-amino-3,4-dimethoxybenzonitrile, benzonitrile, 2-amino-4,5-dimethoxy, 2-amino-4,5-dimethoxy-benzonitrile, 2-amino-4,5-dimethoxybenzenecarbonitrile, pubchem4632, acmc-209wvj
COC1=C(C=C(C(=C1)C#N)N)OC
178.191
178.19
97°C to 101°C
26961-27-3
MFCD00058682
2372149
BJAYMNUBIULRMF-UHFFFAOYSA-N
2-amino-4,5-dimethoxybenzonitrile
735967
98%

Chemical Identifiers

26961-27-3
178.191
BJAYMNUBIULRMF-UHFFFAOYSA-N
735967
COC1=C(C=C(C(=C1)C#N)N)OC
C9H10N2O2
MFCD00058682
6-aminoveratronitrile, 2-cyano-4,5-dimethoxyaniline, 4-amino-5-cyanoveratrole, 6-amino-3,4-dimethoxybenzonitrile, benzonitrile, 2-amino-4,5-dimethoxy, 2-amino-4,5-dimethoxy-benzonitrile, 2-amino-4,5-dimethoxybenzenecarbonitrile, pubchem4632, acmc-209wvj
2-amino-4,5-dimethoxybenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

Documents