Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Amino-4',5'-dimethoxyacetophenone, 98%

$137.12 - $528.88

Chemical Identifiers

Chemical Name or Material 2'-Amino-4',5'-dimethoxyacetophenone
CAS 4101-30-8
Molecular Formula C10H13NO3
Formula Weight 195.22
MDL Number MFCD00016646
Beilstein 391380
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1060506

  SDS
alfa aesar™
L1060506
5g Each for $137.12

AAL1060514

  SDS
alfa aesar™
L1060514
25g Each for $528.88
Description & Specifications

Specifications

Formula Weight 195.22
Health Hazard 1 Warning
MDL Number MFCD00016646
Assay 98%
CAS 4101-30-8
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C10H13NO3
Chemical Name or Material 2'-Amino-4',5'-dimethoxyacetophenone
Melting Point 104° to 108°C
Beilstein 391380
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.