Please login to your online account to display your discounted pricing

2-Amino-4,5-dihydro-1,3-thiazol-4-one hydrochloride, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Amino-4, 5-dihydro-1, 3-thiazol-4-one hydrochloride
CAS 2192-06-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C3H4N2OS·ClH
Formula Weight 152.6
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RH00879DA

  SDS
fisher scientific company
RH00879DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 152.6
Quantity 1g
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
CAS 2192-06-5
Chemical Name or Material 2-Amino-4, 5-dihydro-1, 3-thiazol-4-one hydrochloride
Packaging Amber Glass Bottle
Molecular Formula C3H4N2OS·ClH