Please login to your online account to display your discounted pricing

2-Amino-4,5-dihydro-1,3-thiazol-4-one hydrochloride, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RH00879DA

 View more versions of this product

Catalog No. RH00879DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 152.6
Quantity 1g
Assay 97%
CAS Min % 97.0
Assay Percent Range 97%
CAS 2192-06-5
Chemical Name or Material 2-Amino-4, 5-dihydro-1, 3-thiazol-4-one hydrochloride
Packaging Amber Glass Bottle
Molecular Formula C3H4N2OS·ClH

Specifications

Chemical Name or Material 2-Amino-4, 5-dihydro-1, 3-thiazol-4-one hydrochloride
CAS 2192-06-5
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C3H4N2OS·ClH
Formula Weight 152.6