Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4,5-difluorobenzoic acid, 97%

$46.26 - $542.43

Chemical Identifiers

Chemical Name or Material 2-Amino-4,5-difluorobenzoic acid
CAS 83506-93-8
Molecular Formula C7H5F2NO2
Formula Weight 173.12
MDL Number MFCD00077509
Beilstein 2834162
Synonym 4,5-Difluoroanthranilic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1995003

  SDS
alfa aesar™
L1995003
1g Each for $46.26

AAL1995006

  SDS
alfa aesar™
L1995006
5g Each for $131.84

AAL1995014

  SDS
alfa aesar™
L1995014
25g Each for $542.43
Description & Specifications

Specifications

Formula Weight 173.12
Chemical Name or Material 2-Amino-4,5-difluorobenzoic acid
MDL Number MFCD00077509
CAS 83506-93-8
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 181° to 183°C
Assay 97%
Molecular Formula C7H5F2NO2
Beilstein 2834162
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Synonym 4,5-Difluoroanthranilic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.