Please login to your online account to display your discounted pricing

2-Amino-4,5,6,7-tetrahydro-1-benzo[b]thiophene-3-carboxamide, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 2-Amino-4,5,6,7-tetrahydro-1-benzo[b]thiophene-3-carboxamide
Assay Percent Range 97%
Molecular Formula C9H12N2OS
Formula Weight 196.27
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC450290010

 
thermo fisher scientific
AC450290010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC450290050

 
thermo fisher scientific
AC450290050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.27
Quantity 1g
CAS 4815-28-5
Chemical Name or Material 2-Amino-4,5,6,7-tetrahydro-1-benzo[b]thiophene-3-carboxamide
Assay Percent Range 97%
Molecular Formula C9H12N2OS