missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-(4-methylphenyl)pyrimidine, 96%

$90.05 - $326.32

Chemical Identifiers

CAS 263276-44-4
Molecular Formula C11H11N3
Formula Weight 185.23
MDL Number MFCD00665911
Synonym 4-(p-Tolyl)-2-pyrimidinamine; 2-Amino-4-(p-tolyl)pyrimidine
Chemical Name or Material 2-Amino-4-(4-methylphenyl)pyrimidine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5035703 SDS Alfa Aesar™
H5035703
1g Each for $326.32
AAH50357MD SDS Alfa Aesar™
H50357MD
250mg Each for $90.05
Specifications Description & Specifications

Chemical Identifiers

263276-44-4
185.23
4-(p-Tolyl)-2-pyrimidinamine; 2-Amino-4-(p-tolyl)pyrimidine
C11H11N3
MFCD00665911
2-Amino-4-(4-methylphenyl)pyrimidine

Specifications

263276-44-4
C11H11N3
MFCD00665911
2-Amino-4-(4-methylphenyl)pyrimidine
1g
185.23
4-(p-Tolyl)-2-pyrimidinamine; 2-Amino-4-(p-tolyl)pyrimidine
96%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning