missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-(3-pyridyl)thiazole, 97%

Manufacturer:  Alfa Aesar™ H5863009

 View more versions of this product

Catalog No. AAH5863009



Description

Description

Applications
It is used as a pharmaceutical intermediate.

Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
Specifications

Specifications

2-Amino-4-(3-pyridyl)thiazole
10g
C8H7N3S
4-pyridin-3-yl thiazol-2-amine, 2-amino-4-3-pyridyl thiazole, 4-pyridin-3-yl-thiazol-2-ylamine, 4-pyridin-3-yl-1,3-thiazol-2-amine, 4-3-pyridinyl-2-thiazolamine, 2-thiazolamine, 4-3-pyridinyl, ccris 6699, 2-amino-4-pyridin-3-yl-1,3-thiazole, 4-pyridine-3-yl-thiazol-2-ylamine, 4-3-pyridinyl-1,3-thiazol-2-ylamine
C1=CC(=CN=C1)C2=CSC(=N2)N
177.225
177.22
97%
Brown
30235-27-9
MFCD00218013
XOHZQGAYUHOJPR-UHFFFAOYSA-N
4-pyridin-3-yl-1,3-thiazol-2-amine
154893
Solid

Chemical Identifiers

30235-27-9
177.225
XOHZQGAYUHOJPR-UHFFFAOYSA-N
154893
C1=CC(=CN=C1)C2=CSC(=N2)N
C8H7N3S
MFCD00218013
4-pyridin-3-yl thiazol-2-amine, 2-amino-4-3-pyridyl thiazole, 4-pyridin-3-yl-thiazol-2-ylamine, 4-pyridin-3-yl-1,3-thiazol-2-amine, 4-3-pyridinyl-2-thiazolamine, 2-thiazolamine, 4-3-pyridinyl, ccris 6699, 2-amino-4-pyridin-3-yl-1,3-thiazole, 4-pyridine-3-yl-thiazol-2-ylamine, 4-3-pyridinyl-1,3-thiazol-2-ylamine
4-pyridin-3-yl-1,3-thiazol-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents