Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-(2-thienyl)thiazole, 97%

Chemical Identifiers

Chemical Name or Material 2-Amino-4-(2-thienyl)thiazole
CAS 28989-50-6
Formula Weight 182.25
MDL Number MFCD00457532
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5803704

 
alfa aesar™
H5803704
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5803709

 
alfa aesar™
H5803709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 182.25
Chemical Name or Material 2-Amino-4-(2-thienyl)thiazole
Physical Form Solid
Assay 97%
Color Pale brown
MDL Number MFCD00457532
CAS 28989-50-6
Quantity 2g