missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-4-(2-chloro-3-pyridyl)pyrimidine, 97%

Manufacturer:  Alfa Aesar™ H54957MD

 View more versions of this product

Catalog No. AAH54957MD



Description

Description

Applications
It is used as primary and secondary intermediate.

Notes
Incompatible with strong oxidizing agents, bases, acids. Keep in room temperature.
Specifications

Specifications

2-Amino-4-(2-chloro-3-pyridyl)pyrimidine
870221-49-1
MFCD18157687
2-pyrimidinamine, 4-2-chloro-3-pyridinyl
C1=CC(=C(N=C1)Cl)C2=NC(=NC=C2)N
206.633
206.63
250mg
C9H7ClN4
14484335
DKCVVHMYFZBDAK-UHFFFAOYSA-N
4-(2-chloropyridin-3-yl)pyrimidin-2-amine
53396335
97%

Chemical Identifiers

870221-49-1
206.633
DKCVVHMYFZBDAK-UHFFFAOYSA-N
53396335
C1=CC(=C(N=C1)Cl)C2=NC(=NC=C2)N
C9H7ClN4
MFCD18157687
2-pyrimidinamine, 4-2-chloro-3-pyridinyl
4-(2-chloropyridin-3-yl)pyrimidin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents