missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-4-[2-(Boc-amino)ethyl]thiazole, 97%, Alfa Aesar™

Manufacturer:  Alfa Aesar H63689MD

 View more versions of this product

Catalog No. AAH63689MD



Description

Description

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

2-Amino-4-[2-(Boc-amino)ethyl]thiazole
250mg
C10H17N3O2S
tert-butyl 2-2-aminothiazol-4-yl ethylcarbamate, tert-butyl n-2-2-amino-1,3-thiazol-4-yl ethyl carbamate, 2-amino-4-2-boc-amino ethyl thiazole, tert-butyl 2-2-amino-1,3-thiazol-4-yl ethyl carbamate
CC(C)(C)OC(=O)NCCC1=CSC(=N1)N
243.325
243.33
97%
Brown
956018-34-1
MFCD09998237
IIILZIJRVHFCGL-UHFFFAOYSA-N
tert-butyl N-[2-(2-amino-1,3-thiazol-4-yl)ethyl]carbamate
45926027
Solid

Chemical Identifiers

956018-34-1
243.325
IIILZIJRVHFCGL-UHFFFAOYSA-N
45926027
CC(C)(C)OC(=O)NCCC1=CSC(=N1)N
C10H17N3O2S
MFCD09998237
tert-butyl 2-2-aminothiazol-4-yl ethylcarbamate, tert-butyl n-2-2-amino-1,3-thiazol-4-yl ethyl carbamate, 2-amino-4-2-boc-amino ethyl thiazole, tert-butyl 2-2-amino-1,3-thiazol-4-yl ethyl carbamate
tert-butyl N-[2-(2-amino-1,3-thiazol-4-yl)ethyl]carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

SDS Documents