missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-(trifluoromethyl)pyridine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H54793MD

 View more versions of this product

Catalog No. AAH54793MD



Specifications

Specifications

2-Amino-3-(trifluoromethyl)pyridine-5-boronic acid pinacol ester
250mg
C12H16BF3N2O2
2-amino-3-trifluoromethyl pyridine-5-boronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)N)C(F)(F)F
288.077
288.07
132°C to 134°C
947249-01-6
MFCD12923414
OQZKROYNCVLJKM-UHFFFAOYSA-N
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridin-2-amine
46864103
96%

Chemical Identifiers

947249-01-6
288.077
OQZKROYNCVLJKM-UHFFFAOYSA-N
46864103
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)N)C(F)(F)F
C12H16BF3N2O2
MFCD12923414
2-amino-3-trifluoromethyl pyridine-5-boronic acid pinacol ester
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents