missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-(trifluoromethyl)pyridine-5-boronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H5479303

 View more versions of this product

Catalog No. AAH5479303



Specifications

Specifications

2-Amino-3-(trifluoromethyl)pyridine-5-boronic acid pinacol ester
1g
C12H16BF3N2O2
2-amino-3-trifluoromethyl pyridine-5-boronic acid pinacol ester, 3-trifluoromethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, 5-4,4,5,5-tetra methyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl-pyridin-2-ylamine, 5-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl-3-trifluoromethyl-2-pyridylamine, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl pyridin-2-amine, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl-pyridin-2-yl-amine, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl-pyridin-2-ylamine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-trifluoromethyl pyridin-2-amine, 6-amino-5-trifluoromethyl pyridin-3-yl boronic acid pinacol ester, 6-amino-5-trifluoromethyl pyridine-3-boronic acid pinacol ester
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)N)C(F)(F)F
288.077
288.07
132°C to 134°C
947249-01-6
MFCD12923414
OQZKROYNCVLJKM-UHFFFAOYSA-N
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridin-2-amine
46864103
96%

Chemical Identifiers

947249-01-6
288.077
OQZKROYNCVLJKM-UHFFFAOYSA-N
46864103
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)N)C(F)(F)F
C12H16BF3N2O2
MFCD12923414
2-amino-3-trifluoromethyl pyridine-5-boronic acid pinacol ester, 3-trifluoromethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, 5-4,4,5,5-tetra methyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl-pyridin-2-ylamine, 5-4,4,5,5-tetramethyl 1,3,2-dioxaborolan-2-yl-3-trifluoromethyl-2-pyridylamine, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl pyridin-2-amine, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl-pyridin-2-yl-amine, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3-trifluoromethyl-pyridin-2-ylamine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-trifluoromethyl pyridin-2-amine, 6-amino-5-trifluoromethyl pyridin-3-yl boronic acid pinacol ester, 6-amino-5-trifluoromethyl pyridine-3-boronic acid pinacol ester
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3-(trifluoromethyl)pyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents