Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3-pyridinemethanol, 98%

$227.53 - $1,008.03

Chemical Identifiers

Chemical Name or Material 2-Amino-3-pyridinemethanol
CAS 23612-57-9
Molecular Formula C6H8N2O
Formula Weight 124.14
MDL Number MFCD05663510
Synonym 2-Amino-3-(hydroxymethyl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5001003

  SDS
alfa aesar™
H5001003
1g Each for $227.53

AAH5001006

  SDS
alfa aesar™
H5001006
5g Each for $1,008.03
Description & Specifications

Specifications

Formula Weight 124.14
Chemical Name or Material 2-Amino-3-pyridinemethanol
Quantity 1g
Assay 98%
Molecular Formula C6H8N2O
MDL Number MFCD05663510
CAS 23612-57-9
Synonym 2-Amino-3-(hydroxymethyl)pyridine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.