Please login to your online account to display your discounted pricing

2-amino-3-pyridinecarboxaldehyde 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Amino-3-pyridinecarboxaldehyde
CAS 7521-41-7
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H6N2O
Formula Weight 122.13
MDL Number MFCD01830382
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC380610010

  SDS
thermo fisher scientific
AC380610010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 2 GHS H Statement
Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Assay ≥96.0%
Formula Weight 122.13
Physical Form Powder, Crystals and/or Chunks
Quantity 1g
Color Yellow to light brown
Melting Point 95° to 102°C
CAS Max % 100.0
CAS Min % 96.0
CAS 7521-41-7
Chemical Name or Material 2-Amino-3-pyridinecarboxaldehyde
Assay Percent Range 97%
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 Warning
MDL Number MFCD01830382
Molecular Formula C6H6N2O