missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-3-picoline, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 104385000

 View more versions of this product

Catalog No. AC104385000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Amino-3-picoline
1603-40-3
100.0
111°C
95% min. (GC)
C6H8N2
MFCD00006320
22, V,9, 272
Solubility in water: freely soluble.
CC1=C(N=CC=C1)N
108.14
108.14
96%
95.0
221.0°C to 222.0°C
Authentic
Glass bottle
1.5813 to 1.5833
500g
2-amino-3-picoline, 2-amino-3-methylpyridine, 2-pyridinamine, 3-methyl, 3-methyl-2-pyridinamine, 3-methyl-2-aminopyridine, 3-methyl-2-pyridylamine, 3-picoline, 2-amino, 2-amino-3-methyl pyridine, 2-amino-beta-picoline, pyridine, 2-amino-3-methyl
RGDQRXPEZUNWHX-UHFFFAOYSA-N
3-methylpyridin-2-amine
15347
96%

Chemical Identifiers

1603-40-3
108.14
RGDQRXPEZUNWHX-UHFFFAOYSA-N
15347
CC1=C(N=CC=C1)N
C6H8N2
MFCD00006320
2-amino-3-picoline, 2-amino-3-methylpyridine, 2-pyridinamine, 3-methyl, 3-methyl-2-pyridinamine, 3-methyl-2-aminopyridine, 3-methyl-2-pyridylamine, 3-picoline, 2-amino, 2-amino-3-methyl pyridine, 2-amino-beta-picoline, pyridine, 2-amino-3-methyl
3-methylpyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 216-501-4

RTECSNumber : US1850000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only