Please login to your online account to display your discounted pricing

2-Amino-3-picoline 96%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  104380050

 View more versions of this product

Catalog No. AC104380050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Liquid After Melting
Formula Weight 108.14
Quantity 5g
Color Clear yellow
TSCA TSCA
RTECS Number US1850000
EINECS Number 216-501-4
CAS Min % 95.0
CAS Max % 100.0
Beilstein 22, V,9, 272
Refractive Index 1.5813 to 1.5833 (20°C, 589nm)
Synonym 2-Amino-3-methylpyridine
Packaging Glass bottle
Water 0.8% max.
Chemical Name or Material 2-Amino-3-picoline
MDL Number MFCD00006320
Assay Percent Range 96%
Molecular Formula C6H8N2
Name Note 96%
CAS 1603-40-3
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic in contact with skin.
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Amino-3-picoline
Name Note 96%
CAS 1603-40-3
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C6H8N2
Formula Weight 108.14
MDL Number MFCD00006320
Synonym 2-Amino-3-methylpyridine
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic in contact with skin.

SDS & Safety Documents