missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-3-picoline, 96%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 104381000

 View more versions of this product

Catalog No. AC104381000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Amino-3-picoline
Authentic
1.5813 to 1.5833
221.0°C to 222.0°C
32.0°C to 34.0°C
96%
95.0
95% min. (GC)
MFCD00006320
2-amino-3-methyl pyridine, 2-amino-3-methylpyridine, 2-amino-3-picoline, 2-amino-beta-picoline, 2-pyridinamine, 3-methyl, 3-methyl-2-aminopyridine, 3-methyl-2-pyridinamine, 3-methyl-2-pyridylamine, 3-picoline, 2-amino, pyridine, 2-amino-3-methyl
RGDQRXPEZUNWHX-UHFFFAOYSA-N
3-methylpyridin-2-amine
15347
Liquid After Melting
111°C
Glass bottle
0.8% max.
Yellow
100g
1603-40-3
100.0
C6H8N2
22, V,9, 272
Solubility in water: freely soluble.
CC1=C(N=CC=C1)N
108.14
108.14
96%

Chemical Identifiers

1603-40-3
108.14
RGDQRXPEZUNWHX-UHFFFAOYSA-N
15347
CC1=C(N=CC=C1)N
C6H8N2
MFCD00006320
2-amino-3-methyl pyridine, 2-amino-3-methylpyridine, 2-amino-3-picoline, 2-amino-beta-picoline, 2-pyridinamine, 3-methyl, 3-methyl-2-aminopyridine, 3-methyl-2-pyridinamine, 3-methyl-2-pyridylamine, 3-picoline, 2-amino, pyridine, 2-amino-3-methyl
3-methylpyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic in contact with skin.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 216-501-4

RTECSNumber : US1850000

TSCA : TSCA

SDS Documents
Product Certifications

RUO – Research Use Only