missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-3-nitropyridine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 151120050

 View more versions of this product

Catalog No. AC151120050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Amino-3-nitropyridine
Authentic
Yellow
5g
4214-75-9
100.0
C5H5N3O2
MFCD00006314
2-amino-3-nitro pyridine, 2-amino-3-nitropyridine, 2-amino3-nitropyridine, 2-pyridinamine, 3-nitro, 3-nitro-2-aminopyridine, 3-nitro-2-pyridinamine, 3-nitro-2-pyridylamine, 3-nitro-pyridin-2-ylamine, 3-nitropyridin-2-ylamine, pyridine, 2-amino-3-nitro
BPYHGTCRXDWOIQ-UHFFFAOYSA-N
3-nitropyridin-2-amine
77887
Crystalline Powder and Needles
167°C
Glass bottle
165.9°C to 166.4°C
99%
98.5
98.5% min. (GC)
NH2(C5H3N)NO2
22, I, 631
Solubility in water: 3g/L (20 c)
C1=CC(=C(N=C1)N)[N+](=O)[O-]
139.11
139.11
99%

Chemical Identifiers

4214-75-9
139.11
BPYHGTCRXDWOIQ-UHFFFAOYSA-N
77887
C1=CC(=C(N=C1)N)[N+](=O)[O-]
C5H5N3O2
MFCD00006314
2-amino-3-nitro pyridine, 2-amino-3-nitropyridine, 2-amino3-nitropyridine, 2-pyridinamine, 3-nitro, 3-nitro-2-aminopyridine, 3-nitro-2-pyridinamine, 3-nitro-2-pyridylamine, 3-nitro-pyridin-2-ylamine, 3-nitropyridin-2-ylamine, pyridine, 2-amino-3-nitro
3-nitropyridin-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning

EINECSNumber : 224-144-0

TSCA : TSCA

SDS Documents