missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-3-nitrobenzoic acid, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC443180010

 View more versions of this product

Catalog No. AC443180010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Amino-3-nitrobenzoic acid
204.0°C to 207.0°C
606-18-8
C7H6N2O4
JJPIVRWTAGQTPQ-UHFFFAOYSA-N
182.13
182.13
Glass Bottle
1g
96% min. (HPLC)
2-amino-3-nitro benzoic acid, 2-amino-3-nitro-benzoic acid, 2-amino-3-nitrobenzoic acid, 2-amino-3-nitrobenzonic acid, 2-amino-nitro-benzoesaure, 2-carboxy-6-nitroaniline, 3-nitroanthranilic acid, amino-3-nitrobenzoic acid, benzoic acid, 2-amino-3-nitro, pubchem2233
C1=CC(=C(C(=C1)[N+](=O)[O-])N)C(=O)O
219633
97%

Chemical Identifiers

606-18-8
182.13
2-amino-3-nitro benzoic acid, 2-amino-3-nitro-benzoic acid, 2-amino-3-nitrobenzoic acid, 2-amino-3-nitrobenzonic acid, 2-amino-nitro-benzoesaure, 2-carboxy-6-nitroaniline, 3-nitroanthranilic acid, amino-3-nitrobenzoic acid, benzoic acid, 2-amino-3-nitro, pubchem2233
C1=CC(=C(C(=C1)[N+](=O)[O-])N)C(=O)O
C7H6N2O4
JJPIVRWTAGQTPQ-UHFFFAOYSA-N
219633
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or

GHS Signal Word: Warning

SDS Documents