missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-3'-nitroacetophenone hydrochloride, 98%

Manufacturer:  Alfa Aesar™ A1220506

 View more versions of this product

Catalog No. AAA1220506


 

Specifications

Specifications

2-Amino-3'-nitroacetophenone hydrochloride
210°C to 212°C (decomposition)
5g
MFCD00051997
3-Nitrophenacylamine hydrochloride
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)CN.Cl
216.621
216.62
36765-84-1
98%
C8H9ClN2O3
UN1759
BRFUJSROKAAPJT-UHFFFAOYSA-N
2-amino-1-(3-nitrophenyl)ethanone;hydrochloride
2774374

Chemical Identifiers

36765-84-1
216.621
BRFUJSROKAAPJT-UHFFFAOYSA-N
2774374
C8H9ClN2O3
MFCD00051997
3-Nitrophenacylamine hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

SDS Documents